Learn More
Electron Microscopy Sciences SEM Pin Stub Adapters, 16 mm x 55 mm
SDP
Catalog No. 501929960
Encompass_Preferred
Used to adapt 1/8" (3.1mm) pin diameter SEM stubs. Made from vacuum grade aluminum with a stainless steel set screw. Allen wrench included.
Diameter: 16 mm
Height: 55 mm
Catalog No. 50-192-9960 Supplier Electron Microscopy Sciences Supplier No. 7594007
Used to adapt 1/8" (3.1mm) pin diameter SEM stubs. Made from vacuum grade aluminum with a stainless steel set screw. Allen wrench included.
Diameter: 16 mm
Height: 55 mm