Learn More
Electron Microscopy Sciences SEM Pin Stub Holder 12 3.2 x 8mm Pin Stubs
SDP
Catalog No. 501901516
Encompass
This 62 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to twelve standard 12.7 mm (1/2") pin stubs
Type: 3.2 mm x 8mm Pin
Catalog No. 50-190-1516 Supplier Electron Microscopy Sciences Supplier No. 7595212
This 62 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to twelve standard 12.7 mm (1/2") pin stubs
Type: 3.2 mm x 8mm Pin