Learn More
Electron Microscopy Sciences SEM Stub Tool with Perspex handle
SDP
Catalog No. 501930518
Encompass_Preferred
A useful tool for assisting in loading and unloading SEM pin stubs from a loading stand or specimen box into the microscope. It features a Perspex handle with beryllium copper pins that give the tool a flexible, but stable, grip on the specimen stub.
Catalog No. 50-193-0518 Supplier Electron Microscopy Sciences Supplier No. EMS005
A useful tool for assisting in loading and unloading SEM pin stubs from a loading stand or specimen box into the microscope. It features a Perspex handle with beryllium copper pins that give the tool a flexible, but stable, grip on the specimen stub.