Learn More
Ted Pella Inc Standard SEM Pin Stub Mount, Ø12.7mm x 8mm pin height, 1000/pack
SDP
Catalog No. NC2894811
Encompass_Preferred
FEI, Tescan, ZEISS (also for Philips, Aspex, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC), aluminum, grooved edge
Catalog No. NC2894811 Supplier Ted Pella Inc Supplier No. 161111000
FEI, Tescan, ZEISS (also for Philips, Aspex, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC), aluminum, grooved edge