Learn More
Electron Microscopy Sciences SEM Preparation Stand
SDP
Catalog No. 5028590
Encompass_Preferred
Specimen mounts of different sizes are placed in the outer ring of the holder base. The center hole of the base will hold a bottle of conductive adhesive. One side of the base takes 10 of the 1/8" pin mounts; reverse side takes 10 of 10 to 15 mm diameter mounts.
Catalog No. 50-285-90 Supplier Electron Microscopy Sciences Supplier No. 76750
Specimen mounts of different sizes are placed in the outer ring of the holder base. The center hole of the base will hold a bottle of conductive adhesive. One side of the base takes 10 of the 1/8" pin mounts; reverse side takes 10 of 10 to 15 mm diameter mounts.