Learn More
Electron Microscopy Sciences SEM Sample Holder Set
SDP
Catalog No. 5028690
Encompass_Preferred
For your convenience we now carry a SEM Sample Holder Set to fill all of your needs. The set consists of one universal holder, one vertical mount holder for thin samples, one 3-pin and one 5-pin configuration holder with a key for the set screw all in a finely finished wood box. Please specify the base requirement for your instrument when ordering. Delivery time on this item is approximately 2 weeks.
Catalog No. 50-286-90 Supplier Electron Microscopy Sciences Supplier No. 75930
For your convenience we now carry a SEM Sample Holder Set to fill all of your needs. The set consists of one universal holder, one vertical mount holder for thin samples, one 3-pin and one 5-pin configuration holder with a key for the set screw all in a finely finished wood box. Please specify the base requirement for your instrument when ordering. Delivery time on this item is approximately 2 weeks.