Learn More
Electron Microscopy Sciences SEM Sample Mountant - Dual Use
SDP
Catalog No. 501931182
Encompass_Preferred
This extraordinary mountant can be used as a temporary or permanent bond. The pen is composed of a fiber tip, allowing for even and reproducable application, and may be used in all SEM applications. The glue appears blue, however dries to clear. Free from any solvents, the base of the glue is acrylic. Photo safe and acid free.
Catalog No. 50-193-1182 Supplier Electron Microscopy Sciences Supplier No. 12659
This extraordinary mountant can be used as a temporary or permanent bond. The pen is composed of a fiber tip, allowing for even and reproducable application, and may be used in all SEM applications. The glue appears blue, however dries to clear. Free from any solvents, the base of the glue is acrylic. Photo safe and acid free.