Learn More
Electron Microscopy Sciences SEM Large Single Pin Mount Storage Tube
SDP
Catalog No. 501931251
Encompass_Preferred
A plastic tube and plug ideal for storage or mailing of SEM mounts. The pin, 1/8" (3.1mm) in diameter, is inserted securely to the plug and protected by the tube. Mounts can be up to 38mm diameter. Ideal for storage or mailing. The outer measurements of the tube are 1.57" (40mm) x 2.95" (75mm). Max height of sample on stub is 1.18" (30mm).
I.D.: 39 mm
Size: 40 mm x 75 mm
Sample height: 30 mm
Catalog No. 50-193-1251 Supplier Electron Microscopy Sciences Supplier No. 7653515
A plastic tube and plug ideal for storage or mailing of SEM mounts. The pin, 1/8" (3.1mm) in diameter, is inserted securely to the plug and protected by the tube. Mounts can be up to 38mm diameter. Ideal for storage or mailing. The outer measurements of the tube are 1.57" (40mm) x 2.95" (75mm). Max height of sample on stub is 1.18" (30mm).
I.D.: 39 mm
Size: 40 mm x 75 mm
Sample height: 30 mm