Learn More
Electron Microscopy Sciences SEM Hitachi Single Mount Storage Tube
SDP
Catalog No. 501901650
Encompass_Preferred
A plastic tube and plug for M4 threaded Hitachi mounts. 15 mm in diameter Tube. A 15 mm Hitachi mount can be inserted securely to the plug and then protected by the tube. Ideal for storage as well as mailing. The internal diameter of the tube is 0.83"(21 mm). The Outer measurements of the tube are: 0.9" (23 mm) x2.05” H (52 mm)
Catalog No. 50-190-1650 Supplier Electron Microscopy Sciences Supplier No. 7653110
A plastic tube and plug for M4 threaded Hitachi mounts. 15 mm in diameter Tube. A 15 mm Hitachi mount can be inserted securely to the plug and then protected by the tube. Ideal for storage as well as mailing. The internal diameter of the tube is 0.83"(21 mm). The Outer measurements of the tube are: 0.9" (23 mm) x2.05” H (52 mm)