Learn More
Electron Microscopy Sciences EMS 24 Place SEM Holder
SDP
Catalog No. 501927953
Encompass
This 24-place holder is made from aluminum and offers a stage travel of 100mm x 50mm (which is perfect to be used with the Hitachi VP SEM). The holder has a unique identification number so every hole is identifiable. The holder measures 112.7mm x 63.5mm x 10mm (L x W x H) and has a 3.2mm mounting pin.
Catalog No. 50-192-7953 Supplier Electron Microscopy Sciences Supplier No. 7595867
This 24-place holder is made from aluminum and offers a stage travel of 100mm x 50mm (which is perfect to be used with the Hitachi VP SEM). The holder has a unique identification number so every hole is identifiable. The holder measures 112.7mm x 63.5mm x 10mm (L x W x H) and has a 3.2mm mounting pin.