Learn More
Ted Pella Inc SEM Pin Stub Mount Gripper Tweezers, NM, 30° angle, 117mm long, for 12.7mm pin stubs
SDP
Catalog No. NC9497640
Encompass
Stainless steel gripper tweezers/forceps specially made for handling and transporting grooved SEM pin stub mounts. Sizes available are 12.7, 25 and 32mm (1/2", 1" and 1-1/4") used in FEI/Philips, ZEISS/LEO, Leica, AMRAY, Cambridgde Instruments, ETEC SEMs. Conveniently grasp the grooved head of a scanning electron microscopy pin stub mount.
Catalog No. NC9497640 Supplier Ted Pella Inc Supplier No. 1664
Stainless steel gripper tweezers/forceps specially made for handling and transporting grooved SEM pin stub mounts. Sizes available are 12.7, 25 and 32mm (1/2", 1" and 1-1/4") used in FEI/Philips, ZEISS/LEO, Leica, AMRAY, Cambridgde Instruments, ETEC SEMs. Conveniently grasp the grooved head of a scanning electron microscopy pin stub mount.