Learn More
Electron Microscopy Sciences SEM Specimen Prep-Stand; Multi-Angle
SDP
Catalog No. 501928215
Encompass
A SEM specimen preparation stand with the ability to hold pin mounts on a 45° angle as well as in a vertical and horizontal direction. The stand, which is made from Aluminum, comes complete with screws that are used to secure the mount during preparation. Both 0.5" (12.7 mm) and 1" (25 mm) mounts can be accommodated on the stand. Measures: 2.75" ( 70 mm) x 2" (50 mm) x 1.6" (40 mm).
Catalog No. 50-192-8215 Supplier Electron Microscopy Sciences Supplier No. 7595822
A SEM specimen preparation stand with the ability to hold pin mounts on a 45° angle as well as in a vertical and horizontal direction. The stand, which is made from Aluminum, comes complete with screws that are used to secure the mount during preparation. Both 0.5" (12.7 mm) and 1" (25 mm) mounts can be accommodated on the stand. Measures: 2.75" ( 70 mm) x 2" (50 mm) x 1.6" (40 mm).