Learn More
Electron Microscopy Sciences SEM Stub Vacuum Desiccator
SDP
Catalog No. 5034121
Encompass_Preferred
Precision machined from a block of aluminum anodized black - this Sample Stub Vacuum Desiccators accommodates 18 SEM Mounted Sample Stubs, pin style, under vacuum for year long. Measures: 13 cm x 13cm x 4.5cm High. Chamber cavity is 10.4cm diameter x 18 mm deep. Lid is made with 10 mm thick clear acrylic.
Catalog No. 50-341-21 Supplier Electron Microscopy Sciences Supplier No. 76550
Precision machined from a block of aluminum anodized black - this Sample Stub Vacuum Desiccators accommodates 18 SEM Mounted Sample Stubs, pin style, under vacuum for year long. Measures: 13 cm x 13cm x 4.5cm High. Chamber cavity is 10.4cm diameter x 18 mm deep. Lid is made with 10 mm thick clear acrylic.