Learn More
ECM Bioscience Sema-3A & NRP1/Plexin A1 Receptor Immunocytochemistry Kit, ECM Biosciences
Catalog No. 502068841
Encompass_Preferred
The Sema-3A & NRP1/Plexin A1 receptor kit can be used for immuncytochemical labeling of Sema-3A relative to its receptors, NRP1 and Plexin A1. The kit also includes secondary reagents conjugated to DyLight 488 or DyLight 594 for detection of primary antibodies using green (493Ex/518Em) or red (593Ex/618Em) filter sets. Applications: ICC, IHC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8841 Supplier ECM Bioscience Supplier No. SK7750
The Sema-3A & NRP1/Plexin A1 receptor kit can be used for immuncytochemical labeling of Sema-3A relative to its receptors, NRP1 and Plexin A1. The kit also includes secondary reagents conjugated to DyLight 488 or DyLight 594 for detection of primary antibodies using green (493Ex/518Em) or red (593Ex/618Em) filter sets. Applications: ICC, IHC; Species Reactivity: Hu, Rt, Ms