Learn More
Carbosynth LLC. SEMA3D Blocking Peptide, 100 ug
SDP
Supplier:  Carbosynth LLC. 33R48830.1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
A synthetic peptide for use as a blocking control in assays to test for specificity of SEMA3D antibody, catalog no. 70R-6406
Catalog No. 50-187-0650