Learn More
Aviva Systems Biology Corp SEMA4F Antibody - N-terminal region (ARP46421_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46421_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEMA4F. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human SEMA4F. Alternate names: SEMAM, SEMAW, M-SEMA, PRO2353, m-Sema-MApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59324