Learn More
Aviva Systems Biology Corp SEMA6A Antibody - middle region (ARP49574_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49574_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEMA6A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SEMA6A. Alternate names: HT018, KIAA1368, SEMA, SEMA6A1, SEMAQ, VIA, sema VIaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60117