Learn More
Aviva Systems Biology Corp SEMA6D Antibody - N-terminal region (ARP49584_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49584_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEMA6D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SEMA6D. Alternate names: FLJ11598, KIAA1479ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60119