Learn More
Selleck Chemical LLC Semaglutide 1mg
SDP
Catalog No. 502173497
Encompass_Preferred
Semaglutide (Rybelsus, Ozempic, NN9535, OG217SC, NNC 0113-0217), a long-acting glucagon-like peptide 1 (GLP-1) analogue, is a GLP-1 receptor agonist with the potential for the treatment of type 2 diabetes mellitus (T2DM). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3497 Supplier Selleck Chemical LLC Supplier No. S96971MG
Semaglutide (Rybelsus, Ozempic, NN9535, OG217SC, NNC 0113-0217), a long-acting glucagon-like peptide 1 (GLP-1) analogue, is a GLP-1 receptor agonist with the potential for the treatment of type 2 diabetes mellitus (T2DM). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.