Learn More
Medchemexpress LLC HY-114118 1mg Medchemexpress, Semaglutide CAS:910463-68-2 Purity:>98%
SDP
Catalog No. 502028830
Encompass_Preferred
Medchemexpress, HY-114118 1mg Semaglutide CAS:910463-68-2 Semaglutide, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide has the potential for type 2 diabetes treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8830 Supplier Medchemexpress LLC Supplier No. HY1141181MG
Medchemexpress, HY-114118 1mg Semaglutide CAS:910463-68-2 Semaglutide, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide has the potential for type 2 diabetes treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.