Learn More
AdipoGen Semaglutide (25 mg)
SDP
Supplier:  AdipoGen AGCP30040M025
Encompass_Preferred
Chemical. CAS: 910463-68-2. Formula: C187H291N45O59. MW: 4115.6. Glucagon-like peptide-1 (GLP-1) receptor agonist. Antidiabetic agent against type 2 diabetes (T2D). Stimulates insulin and suppresses glucagon secretion in a glucose-dependent manner. Longer-acting alternative to liraglutide. Chemically similar to glucagon-like peptide-1 (GLP-1), with two amino acid substitutions compared to human GLP-1 (Aib(8), Arg(34)) and derivatized at lysine 26. Antiobesity activity. Reduces energy intake, likely mechanisms for semaglutide-induced weight loss include less appetite and food cravings, better control of eating and lower relative preference for fatty, energy-dense foods.
Catalog No. 50-205-3741