Learn More
Medchemexpress LLC HY-114118 5mg , Semaglutide CAS:910463-68-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1141185MG
Encompass_Preferred
HY-114118 5mg Semaglutide CAS: 910463-68-2 Semaglutide, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide has the potential for type 2 diabetes treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9952