Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Semaglutide (acetate) | 1997361-85-9 | 99.9% | 4113.58 (free base) | 500 UG
SDP

Catalog No. 5000436179
Encompass_Preferred

Semaglutide (acetate) | 1997361-85-9 | 99.9% | 4113.58 (free base) | 500 UG

Catalog No. 50-004-36179 Supplier Medchemexpress LLC Supplier No. HY114118B500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Semaglutide (acetate) | 1997361-85-9 | 99.9% | 4113.58 (free base) | 500 UG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.