Learn More
Medchemexpress LLC Semaglutide acetate | 1997361-85-9 | MFCD03092900 | 99.9% | 4173.6 g/mol | C189H295N45O61 | 5 MG
SDP
Catalog No. 5000264590
Encompass_Preferred
Semaglutide acetate is a long-acting glucagon-like peptide-1 (GLP-1) receptor agonist supplied for research use. The acetate salt form is intended for in-vitro and preclinical studies in metabolic and neurodegenerative disease models (CAS 1997361-85-9). It is provided as a purified peptide reagent for assay development and biological testing.
  • Long-acting GLP-1 receptor agonist for metabolic and neurological research.
  • High purity (reported 99.92%).
  • Supplied in small research pack sizes, including 5 mg.
  • Suitable for in vitro and preclinical pharmacology studies.
  • Acetate salt form for consistent handling and dosing.
Catalog No. 50-002-64590 Supplier Medchemexpress LLC Supplier No. HY114118B5MG
Semaglutide acetate is a long-acting glucagon-like peptide-1 (GLP-1) receptor agonist supplied for research use. The acetate salt form is intended for in-vitro and preclinical studies in metabolic and neurodegenerative disease models (CAS 1997361-85-9). It is provided as a purified peptide reagent for assay development and biological testing.
  • Long-acting GLP-1 receptor agonist for metabolic and neurological research.
  • High purity (reported 99.92%).
  • Supplied in small research pack sizes, including 5 mg.
  • Suitable for in vitro and preclinical pharmacology studies.
  • Acetate salt form for consistent handling and dosing.