Learn More
TARGETMOL CHEMICALS INC Semaglutide Acetate 1MG
SDP
Catalog No. 502878310
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Semaglutide Acetate is an agonist of a glucagon-like peptide 1 (GLP-1) receptor and can be used in studies about the treatment of type 2 diabetes. Purity 98.42%
Catalog No. 50-287-8310 Supplier TARGETMOL CHEMICALS INC Supplier No. T19850L1MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Semaglutide Acetate is an agonist of a glucagon-like peptide 1 (GLP-1) receptor and can be used in studies about the treatment of type 2 diabetes. Purity 98.42%