Learn More
Bachem Semaglutide acetate salt, 910463-68-2, 1 mg
SDP
Supplier:  Bachem 4091661.0001
Encompass_Preferred
Molecular Formula: C₁₈₇H₂₉₁N₄₅O₅₉, Synonyms: NN9535, Relative Molecular Mass: 4113.64, Storage Conditions: -20 ± 5 °C. Semaglutide is under evaluation for the management of diabetes II. Due to its prolonged half-life, the peptide has to be injected only once per week.
Catalog No. NC1495081