Learn More
AdipoGen Semaglutide AcOH
SDP
Supplier:  AdipoGen AGCP30032M005
Encompass_Preferred
Chemical. CAS 910463-68-2 parent. Formula C187H291N45O59 . C2H4O2. MW 4113.6 . 60.0. Synthetic. Glucagon-like peptide-1 GLP-1 receptor agonist. Antidiabetic agent against type 2 diabetes T2D. Stimulates insulin and suppresses glucagon secretion in a glucose-dependent manner. Longer-acting alternative to liraglutide. Chemically similar to glucagon-like peptide-1 GLP-1, with two amino acid substitutions compared to human GLP-1 Aib8, Arg34 and derivatized at lysine 26. Antiobesity activity. Reduces energy intake, likely mechanisms for semaglutide-induced weight loss include less appetite and food cravings, better control of eating and lower relative preference for fatty, energy-dense foods.
Catalog No. 50-256-2425