Learn More
Creative Diagnostics Semaglutide ELISA Kit, 96T, Competitive inhibition enzyme immunoassay, Serum, Plasma, 4-8°C, For research use only
Supplier:  Creative Diagnostics DEIASL092
Encompass
The Creative Diagnostics Semaglutide ELISA Kit is designed for the quantitative measurement of Semaglutide in serum and plasma samples using a highly sensitive competitive inhibition enzyme immunoassay. This kit is particularly useful for researchers studying opioid peptides and type 2 diabetes and obesity. It offers a precise approach for in vitro analysis, ensuring reproducibility and accuracy in results. Here are some key features:
  • Reproducible enzyme immunoassay method
  • Suitable for serum and plasma sample types
  • Storage at 4-8°C for 1 year
  • Highly sensitive detection with thresholds as low as 2 ng/mL
  • Designed for research use only
  • Principle based on competitive immunoassay
  • Includes 96-well immunoplate and essential reagents
Catalog No. NC2179122