Learn More
Medchemexpress LLC Semaphorin-3A/SEMA3A 100ug
SDP
Catalog No. 5000197003
Encompass_Preferred
Recombinant human Semaphorin-3A (SEMA3A) expressed in HEK293 cells and supplied as a lyophilized, purified protein with N-terminal 6xHis and Flag tags. The protein is provided in multiple size options for research use in studies of axon guidance, neuronal development, and SEMA3A signaling. Predicted molecular mass is ~61.3 kDa (apparent 70-75 kDa by SDS-PAGE due to glycosylation). Purity >95% and endotoxin <1 EU/μg; refer to the lot-specific COA for exact buffer and handling information.
  • High purity for consistent experimental results.
  • Low endotoxin suitable for cell-based assays.
  • N-terminal affinity tags facilitate detection and purification.
  • Multiple size options to match experimental scale.
  • Lyophilized format for extended shelf life and easy storage.
  • HEK293 expression preserves mammalian post-translational modifications.
Catalog No. 50-001-97003 Supplier Medchemexpress LLC Supplier No. HYP70797100UG
Recombinant human Semaphorin-3A (SEMA3A) expressed in HEK293 cells and supplied as a lyophilized, purified protein with N-terminal 6xHis and Flag tags. The protein is provided in multiple size options for research use in studies of axon guidance, neuronal development, and SEMA3A signaling. Predicted molecular mass is ~61.3 kDa (apparent 70-75 kDa by SDS-PAGE due to glycosylation). Purity >95% and endotoxin <1 EU/μg; refer to the lot-specific COA for exact buffer and handling information.
  • High purity for consistent experimental results.
  • Low endotoxin suitable for cell-based assays.
  • N-terminal affinity tags facilitate detection and purification.
  • Multiple size options to match experimental scale.
  • Lyophilized format for extended shelf life and easy storage.
  • HEK293 expression preserves mammalian post-translational modifications.