Learn More
Medchemexpress LLC Semaphorin-3A/SEMA3A 10ug
SDP
Catalog No. 5000196557
Encompass_Preferred
Recombinant rat Semaphorin-3A is supplied lyophilized and formulated for stability; it is expressed in E. coli with an N-terminal 6xHis tag and is intended for research on axon guidance and semaphorin signaling.
  • Recombinant rat semaphorin-3a expressed in E. coli with an N-terminal 6xHis tag.
  • Approximately 90.6 kDa molecular weight, >90% purity by reducing SDS-PAGE.
  • Lyophilized formulation containing trehalose to enhance shelf stability.
  • Stable at -20°C for up to two years; reconstituted stability 4°C for 1 week or -20°C for longer with carrier protein.
  • Do not reconstitute below 100 μg/mL in ddH2O to maintain recommended concentration.
  • Intended for in vitro studies of axon guidance and semaphorin signaling pathways.
Catalog No. 50-001-96557 Supplier Medchemexpress LLC Supplier No. HYP7156410UG
Add to Cart
Add to Cart
Recombinant rat Semaphorin-3A is supplied lyophilized and formulated for stability; it is expressed in E. coli with an N-terminal 6xHis tag and is intended for research on axon guidance and semaphorin signaling.
  • Recombinant rat semaphorin-3a expressed in E. coli with an N-terminal 6xHis tag.
  • Approximately 90.6 kDa molecular weight, >90% purity by reducing SDS-PAGE.
  • Lyophilized formulation containing trehalose to enhance shelf stability.
  • Stable at -20°C for up to two years; reconstituted stability 4°C for 1 week or -20°C for longer with carrier protein.
  • Do not reconstitute below 100 μg/mL in ddH2O to maintain recommended concentration.
  • Intended for in vitro studies of axon guidance and semaphorin signaling pathways.