Learn More
Medchemexpress LLC Semaphorin-3A/SEMA3A 20ug | 20ug
SDP
Catalog No. 5000193715
Encompass_Preferred
Semaphorin-3A/SEMA3A Protein guides growth cones and is implicated in sensory projection patterning selectively repelling axons that terminate dorsally It contributes to olfactory system development and neuronal control of puberty SEMA3A interacts with PXND1 although the exact nature of this interaction is unspecified Semaphorin-3A/SEMA3A Protein Mouse (HEK293 His) is the recombinant mouse-derived Semaphorin-3A/SEMA3A protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-93715 Supplier Medchemexpress LLC Supplier No. HYP7835020UG
Add to Cart
Add to Cart
Semaphorin-3A/SEMA3A Protein guides growth cones and is implicated in sensory projection patterning selectively repelling axons that terminate dorsally It contributes to olfactory system development and neuronal control of puberty SEMA3A interacts with PXND1 although the exact nature of this interaction is unspecified Semaphorin-3A/SEMA3A Protein Mouse (HEK293 His) is the recombinant mouse-derived Semaphorin-3A/SEMA3A protein expressed by HEK293 with N-His labeled tag