Learn More
Medchemexpress LLC Semaphorin-3A/SEMA3A protein, human (HEK293, N-His) | 2UG
SDP
Catalog No. 5000177661
Encompass_Preferred
Recombinant human Semaphorin-3A is a secreted guidance protein produced in HEK293 mammalian cells with an N-terminal His tag and supplied as a small research pack (2 μg). The product preserves native glycosylation and folding, exhibits an observed molecular weight of 60-73 kDa by SDS-PAGE, and has low endotoxin levels suitable for many cell-based and biochemical assays. Aliquot and store frozen to maintain activity.
  • Preserves mammalian glycosylation and native folding
  • Low endotoxin suitable for cell-based assays
  • N-terminal His tag simplifies purification and detection
  • Small research pack size ideal for pilot studies
  • Observed molecular weight 60-73 kDa due to glycosylation
  • Stable when aliquoted and stored frozen
Catalog No. 50-001-77661 Supplier Medchemexpress LLC Supplier No. HYP7002972UG
Recombinant human Semaphorin-3A is a secreted guidance protein produced in HEK293 mammalian cells with an N-terminal His tag and supplied as a small research pack (2 μg). The product preserves native glycosylation and folding, exhibits an observed molecular weight of 60-73 kDa by SDS-PAGE, and has low endotoxin levels suitable for many cell-based and biochemical assays. Aliquot and store frozen to maintain activity.
  • Preserves mammalian glycosylation and native folding
  • Low endotoxin suitable for cell-based assays
  • N-terminal His tag simplifies purification and detection
  • Small research pack size ideal for pilot studies
  • Observed molecular weight 60-73 kDa due to glycosylation
  • Stable when aliquoted and stored frozen