Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Semaphorin-3A/SEMA3A 50ug
SDP

Catalog No. 5000196379
Encompass_Preferred

Recombinant rat Semaphorin-3A (SEMA3A) is a secreted axon-guidance protein expressed in E. coli and supplied as an N-terminal His-tagged, lyophilized powder for research use. The material is supplied in 50 μg vials, characterized by >90% purity by reducing SDS-PAGE, has an approximate molecular weight of 90.6 kDa, and is formulated with trehalose for enhanced stability. Store at -20°C; reconstituted material is stable short-term at 4°C or long-term at -20°C with carrier protein.

  • Suitable for biochemical and cell-based assays.
  • High purity (>90%) validated by reducing SDS-PAGE.
  • N-terminal His tag enables affinity capture or detection.
  • Lyophilized formulation with trehalose for improved stability.
  • Recommended storage at -20°C with aliquoting to avoid freeze-thaw.
  • Sequence corresponds to accession Q63548 (N21-V772).

Catalog No. 50-001-96379 Supplier Medchemexpress LLC Supplier No. HYP7156450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat Semaphorin-3A (SEMA3A) is a secreted axon-guidance protein expressed in E. coli and supplied as an N-terminal His-tagged, lyophilized powder for research use. The material is supplied in 50 μg vials, characterized by >90% purity by reducing SDS-PAGE, has an approximate molecular weight of 90.6 kDa, and is formulated with trehalose for enhanced stability. Store at -20°C; reconstituted material is stable short-term at 4°C or long-term at -20°C with carrier protein.

  • Suitable for biochemical and cell-based assays.
  • High purity (>90%) validated by reducing SDS-PAGE.
  • N-terminal His tag enables affinity capture or detection.
  • Lyophilized formulation with trehalose for improved stability.
  • Recommended storage at -20°C with aliquoting to avoid freeze-thaw.
  • Sequence corresponds to accession Q63548 (N21-V772).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.