Learn More
Medchemexpress LLC Semaphorin-3A/SEMA3A 5ug
SDP
Catalog No. 5000195892
Encompass_Preferred
Recombinant rat Semaphorin-3A (SEMA3A) is a His-tagged protein expressed in E. coli corresponding to amino acids N21-V772. Supplied as a 5 μg vial, it is intended for research applications in neuronal development, axon guidance, and cell signaling. Follow supplier documentation for reconstitution and storage instructions.
  • Recombinant rat protein with N-terminal 6xHis tag enabling affinity purification.
  • Amino acid range N21-V772, suitable for functional and structural studies.
  • Expressed in E. coli and supplied in a research-grade format per supplier documentation.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Intended for research use in assays studying axon guidance and neuronal signaling.
Catalog No. 50-001-95892 Supplier Medchemexpress LLC Supplier No. HYP715645UG
Recombinant rat Semaphorin-3A (SEMA3A) is a His-tagged protein expressed in E. coli corresponding to amino acids N21-V772. Supplied as a 5 μg vial, it is intended for research applications in neuronal development, axon guidance, and cell signaling. Follow supplier documentation for reconstitution and storage instructions.
  • Recombinant rat protein with N-terminal 6xHis tag enabling affinity purification.
  • Amino acid range N21-V772, suitable for functional and structural studies.
  • Expressed in E. coli and supplied in a research-grade format per supplier documentation.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Intended for research use in assays studying axon guidance and neuronal signaling.