Learn More
Medchemexpress LLC Semaphorin-3A (SEMA3A) | >95.0% | 50 UG
SDP
Catalog No. 5000292749
Encompass_Preferred
Recombinant mouse Semaphorin-3A (SEMA3A) is a glycosylated, biologically active protein expressed in HEK293 cells and supplied as an N-terminal human Fc-tagged lyophilized powder for in vitro research. The product is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 80-95 kDa, low endotoxin (<1 EU/μg), and is stable when aliquoted and stored frozen for long-term use.
  • Glycosylated, mammalian-expressed protein suitable for activity assays.
  • N-terminal human Fc tag facilitates detection and immobilization.
  • High purity (>95%) verified by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) compatible with cell-based assays.
  • Lyophilized formulation for convenient storage and reconstitution.
  • Stable when aliquoted and stored at -20°C or -80°C for long-term storage.
Catalog No. 50-002-92749 Supplier Medchemexpress LLC Supplier No. HYP7456650UG
Add to Cart
Add to Cart
Recombinant mouse Semaphorin-3A (SEMA3A) is a glycosylated, biologically active protein expressed in HEK293 cells and supplied as an N-terminal human Fc-tagged lyophilized powder for in vitro research. The product is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 80-95 kDa, low endotoxin (<1 EU/μg), and is stable when aliquoted and stored frozen for long-term use.
  • Glycosylated, mammalian-expressed protein suitable for activity assays.
  • N-terminal human Fc tag facilitates detection and immobilization.
  • High purity (>95%) verified by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) compatible with cell-based assays.
  • Lyophilized formulation for convenient storage and reconstitution.
  • Stable when aliquoted and stored at -20°C or -80°C for long-term storage.