Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Semaphorin-3A (SEMA3A) protein | >95.0% | 5 UG
SDP

Catalog No. 5000291813
Encompass_Preferred

Recombinant semaphorin-3A (SEMA3A) is a secreted axon guidance and cell-signaling protein supplied as a research reagent. The product is provided as a small purified preparation intended for in vitro assays, cell biology studies, and biochemical characterization. Purity is reported by the manufacturer as greater than 95% by reducing SDS-PAGE.

  • Recombinant protein suitable for research use only.
  • Expressed in HEK293 cells to support eukaryotic post-translational modifications.
  • Fc-tagged format to aid detection and dimerization where applicable.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Supplied in a small, lyophilized preparation for convenient storage.

Catalog No. 50-002-91813 Supplier Medchemexpress LLC Supplier No. HYP745665UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant semaphorin-3A (SEMA3A) is a secreted axon guidance and cell-signaling protein supplied as a research reagent. The product is provided as a small purified preparation intended for in vitro assays, cell biology studies, and biochemical characterization. Purity is reported by the manufacturer as greater than 95% by reducing SDS-PAGE.

  • Recombinant protein suitable for research use only.
  • Expressed in HEK293 cells to support eukaryotic post-translational modifications.
  • Fc-tagged format to aid detection and dimerization where applicable.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Supplied in a small, lyophilized preparation for convenient storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.