Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Semaphorin-4A/SEMA4A 100ug
SDP

Catalog No. 5000196827
Encompass_Preferred

Recombinant mouse Semaphorin-4A (SEMA4A) ectodomain produced in HEK293 cells with a C-terminal 6xHis tag. The protein participates in axon guidance, synaptic development, and T-cell priming. Supplied lyophilized with formulation buffers for stability, it is characterized by >95% purity and a sequence spanning T33-H682. Store frozen and reconstitute carefully for research applications.

  • Produced in HEK293 mammalian expression system for native post-translational modifications.
  • C-terminal 6xHis tag enables straightforward purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE ensures experimental consistency.
  • Molecular weight range 67-95 kDa accounts for glycosylation variants.
  • Lyophilized formulation with stabilizers improves shelf stability.
  • Recommended reconstitution at a concentration of at least 100 μg/mL in ddH2O to maintain solubility.
  • Store frozen; aliquot for long-term storage at -20 °C or -80 °C.

Catalog No. 50-001-96827 Supplier Medchemexpress LLC Supplier No. HYP71283100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse Semaphorin-4A (SEMA4A) ectodomain produced in HEK293 cells with a C-terminal 6xHis tag. The protein participates in axon guidance, synaptic development, and T-cell priming. Supplied lyophilized with formulation buffers for stability, it is characterized by >95% purity and a sequence spanning T33-H682. Store frozen and reconstitute carefully for research applications.

  • Produced in HEK293 mammalian expression system for native post-translational modifications.
  • C-terminal 6xHis tag enables straightforward purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE ensures experimental consistency.
  • Molecular weight range 67-95 kDa accounts for glycosylation variants.
  • Lyophilized formulation with stabilizers improves shelf stability.
  • Recommended reconstitution at a concentration of at least 100 μg/mL in ddH2O to maintain solubility.
  • Store frozen; aliquot for long-term storage at -20 °C or -80 °C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.