Learn More
Medchemexpress LLC Semaphorin-4A/SEMA4A 10ug
SDP
Catalog No. 5000196434
Encompass_Preferred
Recombinant mouse Semaphorin-4A produced in HEK293 cells with a C-terminal 6xHis tag. The lyophilized protein is supplied in small research quantities (10 μg) at ≥95% purity and is intended for in vitro binding studies, antibody validation, and functional assays; follow reconstitution and storage instructions from the manufacturer.
  • Produced in HEK293 mammalian expression system.
  • C-terminal 6xHis tag for detection and purification.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Reported molecular weight approximately 67-95 kDa.
  • Provided in research-scale quantities suitable for assay development.
  • Store and aliquot at -20°C or -80°C for extended stability.
Catalog No. 50-001-96434 Supplier Medchemexpress LLC Supplier No. HYP7128310UG
Recombinant mouse Semaphorin-4A produced in HEK293 cells with a C-terminal 6xHis tag. The lyophilized protein is supplied in small research quantities (10 μg) at ≥95% purity and is intended for in vitro binding studies, antibody validation, and functional assays; follow reconstitution and storage instructions from the manufacturer.
  • Produced in HEK293 mammalian expression system.
  • C-terminal 6xHis tag for detection and purification.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Reported molecular weight approximately 67-95 kDa.
  • Provided in research-scale quantities suitable for assay development.
  • Store and aliquot at -20°C or -80°C for extended stability.