Learn More
Medchemexpress LLC Semaphorin-4A/SEMA4A 50ug
SDP
Catalog No. 5000196525
Encompass_Preferred
Recombinant mouse Semaphorin-4A extracellular domain expressed in HEK293 cells, supplied lyophilized with a C-terminal 6xHis tag. Provided at 50 μg with >95% purity and formulated for stability; certificate of analysis and datasheet are available for quality verification.
  • Mouse extracellular domain (residues T33-H682).
  • Expressed in HEK293 mammalian cells for proper folding and post-translational modifications.
  • C-terminal 6xHis tag for affinity purification and detection.
  • Lyophilized formulation with stabilizers for extended storage stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Supplied at 50 μg suitable for in vitro research applications.
  • Certificate of analysis and datasheet provided for quality assurance.
Catalog No. 50-001-96525 Supplier Medchemexpress LLC Supplier No. HYP7128350UG
Recombinant mouse Semaphorin-4A extracellular domain expressed in HEK293 cells, supplied lyophilized with a C-terminal 6xHis tag. Provided at 50 μg with >95% purity and formulated for stability; certificate of analysis and datasheet are available for quality verification.
  • Mouse extracellular domain (residues T33-H682).
  • Expressed in HEK293 mammalian cells for proper folding and post-translational modifications.
  • C-terminal 6xHis tag for affinity purification and detection.
  • Lyophilized formulation with stabilizers for extended storage stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Supplied at 50 μg suitable for in vitro research applications.
  • Certificate of analysis and datasheet provided for quality assurance.