Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Semaphorin-4A/SEMA4A 5ug
SDP

Catalog No. 5000196736
Encompass_Preferred

Recombinant mouse Semaphorin-4A is supplied as a His-tagged, HEK293-expressed protein in a lyophilized 5 μg format for laboratory research. It is designed for studies of plexin-mediated signaling and synaptic development, offering high purity and mammalian-like post-translational modifications to support biochemical and cell-based assays.

  • Expressed in HEK293 cells for mammalian-like post-translational modifications.
  • His tag enables straightforward detection and purification.
  • Lyophilized formulation for extended shelf life and easy storage.
  • Greater than 95% purity by reducing SDS-PAGE for reliable results.
  • Molecular weight range 67-95 kDa compatible with SDS-PAGE analysis.
  • Supplied in a 5 μg size appropriate for small-scale experiments.
  • Recommended storage at -20°C or -80°C; aliquot after reconstitution.

Catalog No. 50-001-96736 Supplier Medchemexpress LLC Supplier No. HYP712835UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse Semaphorin-4A is supplied as a His-tagged, HEK293-expressed protein in a lyophilized 5 μg format for laboratory research. It is designed for studies of plexin-mediated signaling and synaptic development, offering high purity and mammalian-like post-translational modifications to support biochemical and cell-based assays.

  • Expressed in HEK293 cells for mammalian-like post-translational modifications.
  • His tag enables straightforward detection and purification.
  • Lyophilized formulation for extended shelf life and easy storage.
  • Greater than 95% purity by reducing SDS-PAGE for reliable results.
  • Molecular weight range 67-95 kDa compatible with SDS-PAGE analysis.
  • Supplied in a 5 μg size appropriate for small-scale experiments.
  • Recommended storage at -20°C or -80°C; aliquot after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.