Learn More
Medchemexpress LLC Semaphorin-4A (SEMA4A) human Fc fusion protein | 100 UG
SDP
Catalog No. 5000292375
Encompass_Preferred
Recombinant human Semaphorin-4A Fc fusion protein comprising the extracellular domain, expressed in HEK293 cells and supplied as a sterile, 0.22 μm-filtered PBS solution (pH 7.4). Intended for in vitro binding, receptor interaction, and cell signaling studies.
  • Human recombinant extracellular domain expressed in HEK293 cells.
  • Fc fusion for improved stability and detection.
  • Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Approximate molecular weight ~110 kDa.
  • Suitable for binding and cell signaling assays.
  • Store at -80°C; ship with dry ice; avoid repeated freeze-thaw cycles.
Catalog No. 50-002-92375 Supplier Medchemexpress LLC Supplier No. HYP76055100UG
Recombinant human Semaphorin-4A Fc fusion protein comprising the extracellular domain, expressed in HEK293 cells and supplied as a sterile, 0.22 μm-filtered PBS solution (pH 7.4). Intended for in vitro binding, receptor interaction, and cell signaling studies.
  • Human recombinant extracellular domain expressed in HEK293 cells.
  • Fc fusion for improved stability and detection.
  • Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Approximate molecular weight ~110 kDa.
  • Suitable for binding and cell signaling assays.
  • Store at -80°C; ship with dry ice; avoid repeated freeze-thaw cycles.