Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Semaphorin-4A/SEMA4A protein, human (HEK293, Fc) | 93.0% | 10 UG
SDP

Catalog No. 5000290807
Encompass_Preferred

Recombinant human Semaphorin-4A Fc fusion protein produced in HEK293 cells and supplied as a sterile, 0.22 μm filtered PBS solution (pH 7.4). The product is provided with analytical data confirming biological activity, an endotoxin specification, and a certificate of analysis reporting high purity. Intended for biochemical and cell-based assays involving SEMA4A interactions and signaling.

  • Expressed in HEK293 cells as an Fc fusion.
  • Supplied as PBS buffer, pH 7.4, 0.22 μm filtered.
  • Endotoxin level <1 EU/μg.
  • Purity 93.0% as determined by reducing SDS-PAGE.
  • Suitable for biochemical and cell-based interaction assays.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-002-90807 Supplier Medchemexpress LLC Supplier No. HYP7605510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Semaphorin-4A Fc fusion protein produced in HEK293 cells and supplied as a sterile, 0.22 μm filtered PBS solution (pH 7.4). The product is provided with analytical data confirming biological activity, an endotoxin specification, and a certificate of analysis reporting high purity. Intended for biochemical and cell-based assays involving SEMA4A interactions and signaling.

  • Expressed in HEK293 cells as an Fc fusion.
  • Supplied as PBS buffer, pH 7.4, 0.22 μm filtered.
  • Endotoxin level <1 EU/μg.
  • Purity 93.0% as determined by reducing SDS-PAGE.
  • Suitable for biochemical and cell-based interaction assays.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.