Learn More
Sino Biological Semaphorin 4D / SEMA4D / CD100 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 80320RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Semaphorin 4D / SEMA4D / CD100 (Catalog#80320-R08H; NP_001164034.1; Met1-Met712). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-7674