Learn More
ACROBiosystems Recombinant Protein;Human Semaphorin 4A / SEMA4A Protein;HEK293;1MG;SEA,SEMA4A,Sema B,SEMAB,SEMB,Semaphorin-4A,Semaphorin-B,Semaphorin 4A
Supplier:  ACROBiosystems SEAH52241MG
Encompass_Preferred
Recombinant Protein;Human Semaphorin 4A / SEMA4A Protein;1MG;Human SEMA4A, His Tag (SEA-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 33 - His 683 (Accession # AAH20974).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.7 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8330