Learn More
ACROBiosystems Recombinant Protein;Human Semaphorin 4D / SEMA4D / CD100 Protein, Fc Tag;HEK293;1MG;CD0,SEMA4D,C9orf164,CD100,FLJ33485,FLJ34282,FLJ39737,FLJ46484,M-sema-G,MGC169138,MGC169141,SEMAJ,coll-4,Semaphorin-4D
Catalog No. 501657503
Encompass_Preferred
Recombinant Protein;Human Semaphorin 4D / SEMA4D / CD100 Protein, Fc Tag;1MG;Human SEMA4D, Fc Tag (CD0-H5257) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Arg 734 (Accession # AAH54500).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 105.4 kDa. The protein migrates as 120-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7503 Supplier ACROBiosystems Supplier No. CD0H52571MG
Recombinant Protein;Human Semaphorin 4D / SEMA4D / CD100 Protein, Fc Tag;1MG;Human SEMA4D, Fc Tag (CD0-H5257) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Arg 734 (Accession # AAH54500).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 105.4 kDa. The protein migrates as 120-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.