Learn More
Medchemexpress LLC HY-15509A 5mg Medchemexpress, Semapimod (tetrahydrochloride) CAS:164301-51-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15509A5MG
Encompass_Preferred
Medchemexpress, HY-15509A 5mg Semapimod (tetrahydrochloride) CAS:164301-51-3 Semapimod tetrahydrochloride (CNI-1493), an inhibitor of proinflammatory cytokine production, can inhibit TNF-α, IL-1β, and IL-6. Semapimod tetrahydrochloride inhibits TLR4 signaling (IC50≈0.3 μM). Semapimod tetrahydrochloride inhibits p38 MAPK and nitric oxide production in macrophages. Semapimod tetrahydrochloride has potential in a variety of inflammatory and autoimmune disorders[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0639