Learn More
Cayman Chemical SematIlIdehydrochlorIde 50mg
SDP
Catalog No. 502941211
Encompass_Preferred
A class III antiarrhythmic agent; prolongs electrically stimulated APD and increases ERP in isolated guinea pig left atria; decreases spontaneous sinoatrial beating rates and induces ERP prolongation of the atrioventricular node in isolated perfused canine hearts; increases ERP, prolonging cardiac refractoriness, in situ in open-chest dog hearts at 1 mg/kg; inhibits PES-induced arrhythmias in dogs
Catalog No. 50-294-1211 Supplier Cayman Chemical Supplier No. 2827250 MG
A class III antiarrhythmic agent; prolongs electrically stimulated APD and increases ERP in isolated guinea pig left atria; decreases spontaneous sinoatrial beating rates and induces ERP prolongation of the atrioventricular node in isolated perfused canine hearts; increases ERP, prolonging cardiac refractoriness, in situ in open-chest dog hearts at 1 mg/kg; inhibits PES-induced arrhythmias in dogs