Learn More
Medchemexpress LLC Semax acetate | 2828433-33-4 | 99.39% | 873.97 | 5 MG
SDP
Catalog No. 5000424745
Encompass_Preferred
Semax acetate is a BBB-penetrable adrenocorticotropic hormone-like peptide that can form stable complexes with Cu2+. It is a synthetic peptide analog of Adrenocorticocorticotropic hormone (ACTH) (4-10). Semax acetate exhibits immunomodulatory, nootropic, and neuroprotective activities and is used in the research of central nervous system diseases such as Alzheimer's disease and cerebral ischemia.
  • BBB-penetrable adrenocorticotropic hormone-like peptide
  • Forms stable complexes with Cu2+
  • Synthetic peptide analog of ACTH (4-10)
  • Possesses immunomodulatory activity
  • Exhibits nootropic activity
  • Demonstrates neuroprotective activity
  • Used in research for central nervous system diseases like Alzheimer's and cerebral ischemia
Catalog No. 50-004-24745 Supplier Medchemexpress LLC Supplier No. HYP1146A5MG
Semax acetate is a BBB-penetrable adrenocorticotropic hormone-like peptide that can form stable complexes with Cu2+. It is a synthetic peptide analog of Adrenocorticocorticotropic hormone (ACTH) (4-10). Semax acetate exhibits immunomodulatory, nootropic, and neuroprotective activities and is used in the research of central nervous system diseases such as Alzheimer's disease and cerebral ischemia.
  • BBB-penetrable adrenocorticotropic hormone-like peptide
  • Forms stable complexes with Cu2+
  • Synthetic peptide analog of ACTH (4-10)
  • Possesses immunomodulatory activity
  • Exhibits nootropic activity
  • Demonstrates neuroprotective activity
  • Used in research for central nervous system diseases like Alzheimer's and cerebral ischemia