Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semen Diluting Fluid, Bicarbonate-Formalin Diluting Fluid for Sperm Count, Ricca Chemical
SDP

Catalog No. 671032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
671032 1 L
6710-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 671032 Supplier Ricca Chemical Company Supplier No. 671032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Semen Diluting Fluid
Color Colorless
Name Note Bicarbonate-Formalin Diluting Fluid for Sperm Count
CAS 7732-18-5, 144-55-8, 50-00-0
CAS Min % 92.0, 4.62, 0.99
CAS Max % 96.0, 4.81, 1.03
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.