Learn More
Cayman Chemical Se-Methylselenocysteine (hydrochloride), Purity: ≥95%, CAS: 863394-07-4, 50 mg
SDP
Catalog No. 502947683
Encompass_Preferred
A selenium-containing amino acid; prevents tert-butyl hydroperoxide-induced increases in MDA levels and glutathione reductase GPX activity, as well as decreases in GSH levels, in HepG2 cells; has an additive effect on irinotecan-induced tumor growth reduction in a FaDu mouse xenograft model at 5 µg/animal per day
Catalog No. 50-294-7683 Supplier Cayman Chemical Supplier No. 3295050 MG
A selenium-containing amino acid; prevents tert-butyl hydroperoxide-induced increases in MDA levels and glutathione reductase GPX activity, as well as decreases in GSH levels, in HepG2 cells; has an additive effect on irinotecan-induced tumor growth reduction in a FaDu mouse xenograft model at 5 µg/animal per day